missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Series High-Performance -20°C Manual Defrost Enzyme Freezers
GREENER_CHOICE

Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers offer a sustainable choice for enzyme storage.

Supplier:  Thermo Scientific™ TSX2320ED

 View more versions of this product

Catalog No. TSX2320ED

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers are designed are designed for applications in which even slight intermittent coil warming during auto defrost cannot be tolerated. The design supports sample protection and sustainability objectives for the storage of enzymes, reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

Every enzyme freezer comes with a complete set of nine shelves with enzyme storage bins to optimize organization and retrieval of smaller materials.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Up to 50% less energy usage than conventional refrigerant models*
  • Natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Temperature range of -35° to -15°C, factory setpoint at -20°C
  • Lower HVAC costs as a result of less heat emission into the environment*
  • Easy to use microprocessor control with setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle
  • Optional chart recorders available
  • Warranty varies by country, please contact us for details
Specifications

Chemical Identifiers

Solid
9
TSX Series
28.5 in.
147.3 cm
Optional
R290
1
Microprocessor
Manual
72.3 cm
Wireless
Smart-Vue Compatible
9

Specifications

Enzyme Freezer
650 L
Solid
9
Microprocessor
TSX Series
Manual
37.5 x 28 x 78.6 in. (95.3 x 71.1 x 199.6 cm)
28.5 in.
72.3 cm
58 in.
147.3 cm
24 in.
61 cm
Optional
15 A
208/230 V
60 Hz
R290
216 kg
191 kg
23 cu. ft.
-25°C to -15°C (-20°C)
1
UL, cUL, cUL
45 Enzyme Bins
Warranty varies by country, please contact us for details
28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)
37.5 in.
95.3 cm
78.6 in.
199.6 cm
28 in.
71.1 cm
Wireless
Smart-Vue Compatible
NEMA 6-15
208-230V, 60Hz
Energy efficient
476 lb.
422 lb.
9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.