missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Series High-Performance -20°C Manual Defrost Enzyme Freezers

Catalog No. p-6962018
Change view
Click to view available options
Capacity (English):
23 cu. ft.
29.2 cu. ft.
Plug Type:
NEMA 5-15
NEMA 6-15
Electrical Requirements:
115V, 60Hz
208-230V, 60Hz
Type:
Enzyme Freezer
High-Performance Freezer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Plug Type Electrical Requirements Type
TSX2320EA 23 cu. ft. NEMA 5-15 115V, 60Hz Enzyme Freezer
TSX3020EA 29.2 cu. ft. NEMA 5-15 115V, 60Hz High-Performance Freezer
TSX3020ED 29.2 cu. ft. NEMA 6-15 208-230V, 60Hz High-Performance Freezer
TSX2320ED 23 cu. ft. NEMA 6-15 208-230V, 60Hz Enzyme Freezer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. TSX2320EA Supplier Thermo Scientific™ Supplier No. TSX2320EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers offer a sustainable choice for enzyme storage.

Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers are designed are designed for applications in which even slight intermittent coil warming during auto defrost cannot be tolerated. The design supports sample protection and sustainability objectives for the storage of enzymes, reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

Every enzyme freezer comes with a complete set of nine shelves with enzyme storage bins to optimize organization and retrieval of smaller materials.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Up to 50% less energy usage than conventional refrigerant models*
  • Natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Temperature range of -35° to -15°C, factory setpoint at -20°C
  • Lower HVAC costs as a result of less heat emission into the environment*
  • Easy to use microprocessor control with setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle
  • Optional chart recorders available
  • Warranty varies by country, please contact us for details

Specifications

Type Enzyme Freezer
Capacity (English) 23 cu. ft.
Capacity (Metric) 650 L
Temperature (Setpoint) -25°C to -15°C (-20°C)
Door Style Solid
No. of Doors 1
Shelves 9
Certifications/Compliance UL, cUL, cUL
Display Microprocessor
Includes 45 Enzyme Bins
Product Line TSX Series
Warranty Warranty varies by country, please contact us for details
Defrost Manual
Dimensions (D x W x H) Interior 28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)
Dimensions (L x W x H) Exterior 37.5 x 28 x 78.6 in. (95.3 x 71.1 x 199.6 cm)
Depth (English) Exterior 37.5 in.
Depth (English) Interior 28.5 in.
Depth (Metric) Exterior 95.3 cm
Depth (Metric) Interior 72.3 cm
Height (English) Exterior 78.6 in.
Height (English) Interior 58 in.
Height (Metric) Exterior 199.6 cm
Height (Metric) Interior 147.3 cm
Width (English) Exterior 28 in.
Width (English) Interior 24 in.
Width (Metric) Exterior 71.1 cm
Width (Metric) Interior 61 cm
Monitoring Options Wireless
Chart Recorder Optional
Smart View Compatibility Smart-Vue Compatible
Amperage 15 A
Plug Type NEMA 5-15
Voltage 115 V
Electrical Requirements 115V, 60Hz
Frequency 60 Hz
Refrigerant R290
Shipping Weight (English) 497 lb.
Shipping Weight (Metric) 225 kg
Weight (English) 443 lb.
Weight (Metric) 201 kg
Rack Capacity 9
Show More Show Less

*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.