Learn More
Fisherbrand™ Superfrost™ Cytogenics Slides
Catalog No. 22035900
Change view
Click to view available options
25 x 75 mm (1 x 3 in.); 1mm thick
Catalog No.
22035900 25 x 75 mm (1 x 3 in.); 1mm thick
1 options
Catalog No. 22035900 Supplier Fisherbrand™ Supplier No. CGN4951600621
Only null left
Specifically processed glass surface inspected for cytogenetic applications
Specifically processed, inspected glass surface.
  • Consistent, high-quality surface for reliable chromosome spreads
  • Hydrophilic surface improves retention of metaphase cells
  • Superfrost white tab for permanent marking
Specifications
Type Microscope Slide
No. per Pack 144