Learn More
Thermo Scientific™ SUN-SRi™ Luer-Slip Syringes
Siliconized and assembled with scale and tip shield.
$417.43 - $983.94
Products 2
Catalog Number Mfr. No. Volume (Metric) Product Type Sterility Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Product Type Sterility Price Quantity  
22-314384
View Documents Promotion Details
Thermo Scientific™
400008
5 mL Luer-slip Syringe Non-sterile
Case of 1000 Each for $983.94
Only null left
22-314383
View Documents Promotion Details
Thermo Scientific™
400004
2 mL Luer-slip Syringe Non-sterile
Case of 1000 Each for $417.43
Only null left
Description
Description
Syringe, Chemically resistant, inert polypropylene and polyethylene, Luer Slip, Bulk
  • Chemically resistant, inert polypropylene and polyethylene
  • No rubber plunger seals or silicone lubricants that may cause sample contamination
  • Bulk packaged, choice or sterile or nonsterile
  • LUER-Slip connection
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'