Learn More
Thermo Scientific™ SUN-SRi™ Luer-Slip Syringes
Siliconized and assembled with scale and tip shield.
Supplier: Thermo Scientific™ 400004
Description
Syringe, Chemically resistant, inert polypropylene and polyethylene, Luer Slip, Bulk
- Chemically resistant, inert polypropylene and polyethylene
- No rubber plunger seals or silicone lubricants that may cause sample contamination
- Bulk packaged, choice or sterile or nonsterile
- LUER-Slip connection
Specifications
| Plastic | |
| Single-use | |
| Centered | |
| Non-sterile |
| Luer-slip Syringe | |
| 2 mL | |
| Leur Slip |