Learn More
Thermo Scientific™ Sorvall™ Culture Flask Adapters
For processing Nunc™ T-75 or T-25 EasyFlasks™ in Thermo Scientific Sorvall Legend™ T/RT Centrifuges.
$2270.25 - $2291.85
Specifications
| For Use With (Equipment) | Thermo Scientific™ Sorvall™ Legend™ T/RT rotor with round bucket |
|---|
Description
Unique angled design ensures tight pelletization, allowing easy removal of supernatant without pellet disruption.
- T-25 adapter holds flask at 25° angle; T-75 at 75°
- Sold in pairs
Specifications
| Thermo Scientific™ Sorvall™ Legend™ T/RT rotor with round bucket |