Learn More
Thermo Scientific™ Sorvall™ Culture Flask Adapters
Catalog No. 75008384
Change view
Click to view available options
Tube Type:
T-25 Flask
T-75 Flask
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tube Type
75008384 T-25 Flask
75008383 T-75 Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75008384 Supplier Thermo Scientific™ Supplier No. 75008384
Requires
Rotor 75006445 with round bucket 75006441
For processing Nunc™ T-75 or T-25 EasyFlasks™ in Thermo Scientific Sorvall Legend™ T/RT Centrifuges.
Unique angled design ensures tight pelletization, allowing easy removal of supernatant without pellet disruption.
  • T-25 adapter holds flask at 25° angle; T-75 at 75°
  • Sold in pairs
Specifications
Tube Type T-25 Flask
For Use With (Equipment) Thermo Scientific™ Sorvall™ Legend™ T/RT rotor with round bucket
Angle 25°