Description
- Ready-to-use mucoliquefying preservative
- Liquefies (with agitation) mucoid cytology specimens
- Maintains cellular detail
- Short-term preservation of specimens
- Histology
- Cytology
Specifications
| Light Red | |
| 4 x 3.8L (1 gal.) | |
| Liquid |
| Tissue Preservative | |
| Provides short term preservation of specimens |