Learn More
Epredia™ Shandon™ Plastic Coverplates
For use on Shandon Sequenza™ and Cadenza™ Immunostainers
$482.86 - $5730.97
Specifications
Product Type Coverplate
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
28-602-18
View Documents
Epredia™
72110013
250/Cs.
Case of 250 Each for $5,730.97
Only null left
72-110-017
View Documents
Epredia™
72110017
25/Pk.
Case of 25 Each for $482.86
Only null left
Description
Description
Saves on the use of costly reagents.
  • Device that fits over standard microscope slide allowing specimens to be immunostained with a minimum amount of reagent
  • Frees considerable technician time
  • Eliminates handling of slides between steps
  • Allows sections to be placed virtually anywhere on slide
  • Protects specimen from drying during long incubations
  • Ensures consistency in staining quality
Specifications
Specifications
Coverplate
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'