Learn More
Epredia™ Shandon™ Plastic Coverplates
Catalog No. 2860218
Change view
Click to view available options
Quantity:
25/Pk.
250/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2860218 250/Cs.
72110017 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2860218 Supplier Epredia™ Supplier No. 72110013
For use on Shandon Sequenza™ and Cadenza™ Immunostainers
Saves on the use of costly reagents.
  • Device that fits over standard microscope slide allowing specimens to be immunostained with a minimum amount of reagent
  • Frees considerable technician time
  • Eliminates handling of slides between steps
  • Allows sections to be placed virtually anywhere on slide
  • Protects specimen from drying during long incubations
  • Ensures consistency in staining quality
Specifications
Product Type Coverplate
Quantity 250/Cs.