missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMB Substrate Solution

Catalog No. ENN301
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENN301 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENN301 Supplier Thermo Scientific™ Supplier No. N301
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A ready-to-use substrate system containing 3.3', 5.5'-tetramethylbenzidine (TMB)

 

  • Ready to use, no preparation required
  • Validated for use with Thermo Scientific Pierce Matched Antibody Pairs and MiniKits
  • Size: 100 mL (sufficient for about ten 96-well microplates)

 

Specifications

Product Line Pierce
Product Type TMB Substrate
Substrate Properties Chemical Substrate
Content And Storage Store between 2°C and 8°C.
Detection Method Colorimetric
Substrate Type HRP (Horseradish Peroxidase) Substrate
Quantity 100 mL

For Research Use Only. Not for use in diagnostic procedures.