missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Poly HRP Dilution Buffer

Catalog No. ENN501
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENN501 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENN501 Supplier Thermo Scientific™ Supplier No. N501
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifically designed to optimize performance of Poly-HRP Streptavidin

Thermo Scientific Pierce Poly-HRP Dilution Buffer is optimized to ensure optimal performance with Poly-HRP Streptavidin.

Features of Poly-HRP Dilution Buffer:

• Formulation: 1% biotin-free casein in a PBS buffer solution.
• Application: Dilution of Poly-HRP Streptavidin (Product # N200): Gently invert bottle and swirl to mix before use. Do not vortex. Use at full strength. Refer to Poly-HRP Streptavidin Certificate of Analysis for recommended dilution ranges. Dilute Poly-HRP Streptavidin immediately before use.

Related Products
Poly-HRP Streptavidin

Specifications

Chemical Name or Material Dilution Buffer
Physical Form Liquid
Quantity 1 L

For Research Use Only. Not for use in diagnostic procedures.