missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Poly-HRP Dilution Buffer

Catalog No. ENN500
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENN500 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENN500 Supplier Thermo Scientific™ Supplier No. N500
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifically designed to optimize performance of Poly-HRP Streptavidin

  • 1% biotin-free casein in a PBS buffer solution
  • Ready-to-use

Specifications

Chemical Name or Material Dilution Buffer
Physical Form Liquid
Quantity 100 mL

For Research Use Only. Not for use in diagnostic procedures.