missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Firefly Luc One-Step Glow Assay Kit

Catalog No. PI16196
Change view
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI16196 100 Reaction Kit
PI16197 1000 Reaction Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI16196 Supplier Thermo Scientific™ Supplier No. 16196

Perform firefly luciferase assays in high-throughput screening applications.

The Thermo Scientific Pierce Firefly One-Step Luc Glow Assay Kit measures firefly luciferase activity in mammalian cell culture using a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications.

This glow-type luciferase assay kit is designed for use with cultured mammalian cells that are transfected with vectors expressing the appropriate firefly luciferase reporter enzyme. Several features make the kit especially suitable for luminometers without injectors and for high-throughput screening (HTS) applications. The convenient, one-step, homogenous protocol minimizes handling steps to support the use of automation. Simply combine the supplied substrate solution and assay buffer to make a single working solution, and then add it to microplate wells containing the transfected, treated cells. The stable signal output (glow) of this system provides flexibility with regard to incubation time. When used with Thermo Scientific Firefly Luc Plasmids, the kit provides a highly sensitive bioluminescent reporter assay system for the detection of promoter or pathway activity.

Highlights

  • Sensitive—highly sensitive detection of firefly luciferase activity
  • Convenient—one-step, mix-and-read protocol; no additional reagents required
  • Stable—increased signal half-life and stability compared to flash assays
  • Simple—does not require a luminometer with injectors
  • Automation-friendly—amenable to high throughput assays

Requires

Firefly luciferase and luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers

Applications

  • Promoter studies for analyzing cis- and trans-acting regulatory elements
  • Drug screening
  • siRNA and miRNA screening
  • Multiplexed assays to study off-target effects
  • Protein localization reporter assays
  • Signal transduction pathway analysis

Product details

Firefly luciferase is a 62 kDa protein produced in nature by several species of the Lampyridae family of beetles, which includes the genera Photinus and Luciola. Thermo Scientific Red Firefly Luciferase has an emission range of 560-700 nm (λmax = 613 nm). Bioluminescent signal from firefly luciferase originates from the oxidation of D-Luciferin. Light output captured using a luminometer can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly luciferase expression. Firefly luciferase bioluminescence provides researchers with a non-radioactive assay for measuring transcriptional activity of regulatory elements. Firefly luciferase reporter activity can be used to determine promoter activity, to study upstream pathway activity in the cell, or to determine effect of activators or inhibitors.

Specifications

Type Assay Kit
Target Luciferase, Firefly Luciferase, Cypridina Luciferase
Sufficient For 100 Luciferase Assays in Microplate Wells
Quantity 100 Reaction Kit
Content And Storage • Firefly Luc One-Step Substrate, Lyophilized (makes 1μL of 1X), store at -20°C
• Firefly Luc One-Step Assay Buffer, 11 mL, store at -20°C
Upon receipt store kit at -20°C.
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Substrate D-Luciferin
Show More Show Less
I prepared my Luciferase Glow Assay Working Solution four hours ago, can I still use it?

All Luciferase Glow Assay Working Solutions should be stored protected from light.

  • Renilla Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for up to 8 hours at room temperature.
  • Gaussia Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for up to 4 hours at room temperature. 
  • Cypridina Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for up to 2 hours at room temperature.
  • Firefly Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for 4 hours at room temperature. For long term use, store at -20 degrees C for up to 2 months.

  • What is the benefit of using the TurboLuc Luciferase One-Step Glow Assay Kit over the Luciferase Glow Assay kits?

    TurboLuc Luciferase One-Step Glow Assay Kit measures activity in mammalian cells with a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications. Several features of the TurboLuc Luciferase One-Step Glow Assay Kit make it suitable for luminometers without injectors and for HTS applications. The convenient, one-step, homogenous protocol minimizes handling steps to support the use of automation. Simply combine the supplied substrate solution and assay buffer to make a single working solution, and then add it to microplate containing the transfected, treated cells. The stable signal output (glow enzyme kinetics) of this system provides flexibility with regard to lead time before read. When used with Thermo Scientific TurboLuc 16 Luciferase vectors, the TurboLuc Luciferase One-Step Glow Assay Kit provides a highly sensitive bioluminescent reporter assay system for the detection of promoter or pathway activity.

    For Research Use Only. Not for use in diagnostic procedures.