missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Everylab Liquid Handling Package
Description
Bring your molecular work together by using the Every Lab Package
Covers basic and complex workflows
- Our ventilated and refrigerated microcentrifuges reduce spin times by up to 20%, making them ideal for high-end molecular or genomic research applications
- The Thermo Scientific™ included 24 X 1.5/2.0mL rotor with ClickSeal™ biocontainment lid allows you to run tubes from 1.5 to 2.0mL tube to mini-preps and spin columns
- Pipettes cover a range of volumes to match necessary volumes, from 0.2-2μL to 100-1000μL
- Snap Cap tubes offer versatility in the lab: Can be frozen or heated in a water bath
Save time with faster sample processing
- Lightweight rotors provide acceleration up to 21,100 x g in 12 seconds.
- Refrigerated models offer cooling from room temperature to set temperature in just 9 minutes.
- Color coding of pipettes helps to select the correct pipette volume while working
Ideal for everyday laboratory use
- High performance engineered polymer rotors are designed for outstanding chemical resistance to minimize corrosion from aggressive reagents
- Conforms to the latest clinical and safety standards including 98/79/EC In Vitro Diagnostic Directive
- Pipettes are fully autoclavable, minimizing disruption and downtime
- Finnpipette pipettes are made of polyvinylidene fluoride (PVDF) components that stand up to harsh chemical, UV light and physical damage
Specifications
Specifications
| Capacity | 24 x 1.5/2 mL |
| Max. Speed | 14,800 rpm |
| Max. RCF | 21,100 x g |
| Includes | Part numbers in package: 05-719-511 (F2 GLP Pipette Kit) and 21-402-902 (1.5 mL microcentrifuge tubes). Note: 01-185-012 or 01-189-180 (centrifuge) ordered separately includes: 75002436 (ventilated) or 75002446 (refrigerated) |
| Refrigerated | Yes |
| Product Line | Thermo Scientific |
| Voltage | 120 V |