missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Owl™ Glove Box Stands

Protecting researchers from direct contact with experiments

$249.78

Specifications

Product Type Stand
For Use With (Application) Electrophoresis System
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (Metric) Max. Load Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Max. Load Price Quantity & Availability  
OWMP01
View Documents Promotion Details
Fisherbrand™
MP01
Single
Each for $249.78
Only null left
 
Description

Description

Specifications

Specifications

Stand
Electrophoresis System
Videos
SDS
Documents

Documents