missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters
Description
Use Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards with Orion™ AQUAfast Colorimeters such as AQ4000 Advanced Colorimeter and Orion AQUAfast II AQ2040 Colorimeter. Orion colorimeters, which measure COD chemistries, are ideal for environmental and wastewater monitoring. Use these calibration standards to check operations prior to using the instruments.
- For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ2040
- Two Calibration Standards: 1000 ppm and 10000ppm, both in 475 mL bottles
Order Info
Reagents sold separately.
Specifications
Specifications
| Concentration | 1000 ppm |
| Certifications/Compliance | EPA |
| For Use With (Equipment) | AQUAfast Colorimeter |
| Industry Type | Environmental Testing, Wastewater Monitoring |
| Quantity | 475 mL |