missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TopVision Agarose Tablets

Catalog No. FERR2802
Change view
Click to view available options
Quantity:
1000 Tablets (0.5 g each)
200 Tablets (0.5 g each)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR2802 1000 Tablets (0.5 g each)
FERR2801 200 Tablets (0.5 g each)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERR2802 Supplier Thermo Scientific™ Supplier No. R2802

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality, multipurpose agarose tablets in blister packaging.

Description
TopVision Agarose Tablets, low EEO (LE), is multipurpose agarose in tablet format delivered in a convenient blister pack. The agarose tablets are made of standard melting point agarose that yields high resolution, sharp DNA bands with high clarity and low background. Its optimized gel strength enhances ease of gel processing and handling. Manufactured using an innovative Organic Solvent-Free manufacturing process that is greener and more environmentally friendly.
Features
• Convenient tablet format - no messy weighing required.
• Fast dissolving protocol.
•“Green”- manufactured with significantly reduced environmental impact, using an innovative organic solvent-free technology.
• Enhanced resolution and reduced background.
Applications
Ideal for routine DNA and RNA gel electrophoresis and blotting assays.
Environmentally responsible Green Solution
Unlike traditional agarose powder, which production relies heavily on the use of toxic organic solvents, TopVision Agarose Tablets are manufactured using an innovative process that completely eliminates dependency on organic solvents and significantly reduces environmental impact while creating a safer working environment.

Appearance:White to off-white powder
> Ash:≤ 0.5%
> CAS:9012-36-6
> EEO:≤ 0.13
> Gel Strength:≥ 1200 g/cm2 (1% Gel)
> Gelling Point:36°C ± 1.5°C (1.5% gel)
> Hazardous:No
> Impurities (DNAses, RNases and Proteases):None
> Melting Point:88°C ± 1.5°C (1.5% gel)
> Moisture:≤ 10%
> Shelf Life:24 Months
> Shipping Condition:Ambient
> Solubility:Clear colorless solution at 1g in 100 mL water
> Storage Condition:Ambient
> Sulfate:≤ 0.15%

Specifications

Separation Range 100 to >30000 bp
Melting Point 88 °C
Quantity 1000 Tablets (0.5 g each)
Validated Application Gel Electrophoresis
Product Line TopVision
Product Type Top Vision Agarose Tablet
How long does it take to dissolve a Thermo Scientific TopVision Agarose Tablet?

Add the tablet into the determined amount of buffer, presoak for two minutes, and proceed the same way as you are used to with the traditional agarose powder. Using tablets saves 15-20 minutes hands-on-time.

At what voltage should I run an agarose gel prepared from TopVision Agarose Tablets?

The electrophoresis conditions do not depend on the form of delivery of agarose. Whether it is loose powder or tablets, the material has the same physicochemical conditions. In general, 4-10 volts/cm (cm is determined by measuring the interelectrode distance, not the gel length) are recommended when running agarose gels under normal horizontal electrophoretic conditions.

Do TopVision Agarose Tablets perform the same compared to traditional agarose powder products?

Yes, the performance is the same, the convenience is much greater.The critical characteristics of agaroses (melting/gelling temperature, gel strength, electroendosmosis (EEO)) impacting the performance are the same.

What is the advantage of using agarose tablets rather than loose powder agarose?

A tablet is a more convenient format, eliminating the messy and time-consuming weighing step.

For Research Use Only. Not for use in diagnostic procedures.