missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ NHS-PEG Solid-Phase Biotinylation Kit - Mini-Spin Columns

Catalog No. PI21450
Change view
Click to view available options
Quantity:
8 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21450 8 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21450 Supplier Thermo Scientific™ Supplier No. 21450

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Kit with Prepacked format (1 to 10mg IgG): Immobilized Nickel Chelate Column 1mL, BupH Phosphate Buffered Saline 1 pack (makes 500mL), No-Weigh NHS-PEG4-Biotin Microtubes 8 x 2mg, 4M Imidazole Stock Solution 5mL
  • Kit with Mini-Spin format (0.1 to 1mg IgG): Nickel Chelated SwellGel Discs (pack of 10), Mini-Spin columns (pack of 10), 2mL Microcentrifuge tubes (pack of 30), BupH Phosphate Buffered Saline 1 pack (makes 500mL), No-Weigh NHS-PEG4-Biotin Microtubes 8 x 2mg, 4M Imidazole Stock Solution 5mL

A column-based approach to antibody biotinylation and clean-up

This innovative antibody-labeling system uses nickel-chelated agarose to temporarily immobilize antibody molecules via their histidine-rich Fc regions. Once held in place on the resin, the antibody can be biotinylated at either sulfhydryl groups (after mild reduction or disulfide bonds) or primary amines. Excess labeling reagent and byproducts are then washed away before recovering the labeled and purified antibody from the resin using a mild imidazole solution. No gel filtration or dialysis is needed. Four kits are available for small (0.1-1mg) or large (1-10mg) antibody samples using either amine-directed (NHS ester) or sulfhydryl-directed (maleimide) labeling reagents.

Highlights:

  • Fast labeling and purification - the entire procedure takes only one hour (two hours for sulfhydryl labeling kits)
  • Easy removal of spent and excess labeling reagent - simply wash away the reaction byproducts - no need for dialysis or gel filtration
  • No dilution effects - solid-phase method allows initially dilute antibodies to be recovered in a smaller volume after labeling
  • Optimized protocols - specific protocols for antibody ensure appropriate level of labeling (2-5 biotins per antibody molecule), minimizing possibility of inactivation caused by overlabeling
  • Sufficient reagents for eight biotin-labeling experiments - Thermo Scientific No-Weigh Packaging ensures that the biotin reagent is fully active for eight separate experiments

Specifications

Product Type NHS-PEG Solid-Phase Biotinylation Kit
Quantity 8 reactions
Content And Storage Sufficient For: 8 labeling reactions, each with 0.1 to 1 mg of antibody
• Ni-NTA Spin Columns, 0.2 mL, 8 columns
• PBS pack (makes 500 mL), 1 pack
• NHS-PEG4-Biotin, 8 x 2 mg
• Imidazole Stock Solution (4M), 5 mL
• Collection Tubes (2mL), 30 tubes

Store at 4°C.
Labeling Scale 0.5 to 1 mg
Product Line EZ-Link
Label or Dye Biotin
Spacer Pegylated
Labeling Target IgG
How do the EZ-Link Micro Biotinylation Kits differ from the standard biotinylation kits?

The standard biotinylation kits contain components for labeling 1-10 mg of protein, and the Micro Biotinylation Kits are structured for labeling 50-200 µg of protein in 200-700 mL. The Micro kits contain the biotinylation reagents in small, individual packages and do not include components to perform the HABA assay.

Can you provide the shelf-life for EZ-Link NHS-PEG Solid-Phase Biotinylation Kit?

EZ-Link NHS-PEG Solid-Phase Biotinylation Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.