missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Every Lab Package

Catalog No. p-10022134
Change view
Click to view available options
Product Line:
Sorvall
Refrigerated:
No
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line Refrigerated
01185012 Sorvall No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01185012 Supplier Thermo Scientific™ Supplier No. 75002439
Only null left

Bring your molecular work together by using the Every Lab Package.

The Everylab Microcentrifuge and Liquid Handling package provides a convenient offering that includes a microcentrifuge, Finnpipettes F2 GLP Kit (includes 4 single channel pipettes, stand, and tips), and a case of 1.5 mL microcentrifuge tubes (5000 tubes).

Covers basic and complex workflows 

  • Our ventilated and refrigerated microcentrifuges reduce spin times by up to 20%, making them ideal for high-end molecular or genomic research applications
  • The Thermo Scientific™ included 24 X 1.5/2.0mL rotor with ClickSeal™ biocontainment lid allows you to run tubes from 1.5 to 2.0mL tube to mini-preps and spin columns 
  • Pipettes cover a range of volumes to match necessary volumes, from 0.2-2μL to 100-1000μL 
  • Snap Cap tubes offer versatility in the lab: Can be frozen or heated in a water bath

Save time with faster sample processing  

  • Lightweight rotors provide acceleration up to 21,100 x g in 12 seconds.
  • Refrigerated models offer cooling from room temperature to set temperature in just 9 minutes. 
  • Color coding of pipettes helps to select the correct pipette volume while working

Ideal for everyday laboratory use 

  • High performance engineered polymer rotors are designed for outstanding chemical resistance to minimize corrosion from aggressive reagents
  • Conforms to the latest clinical and safety standards including 98/79/EC In Vitro Diagnostic Directive
  • Pipettes are fully autoclavable, minimizing disruption and downtime
  • Finnpipette pipettes are made of polyvinylidene fluoride (PVDF) components that stand up to harsh chemical, UV light and physical damage

Specifications

Capacity 24 x 1.5/2mL
Max. Speed 14,800 rpm
Max. RCF 21,100 x g
Type Microcentrifuge
Includes
  • Everylab Centrifuge Ventilated Package - Sorvall Legend 120 V, 21 V (including 24 x 1.5/2 mL rotor) 
  • F2 Single Channel Pipettes - GLP Kit 2 
  • Snap Cap tubes (1 case 5000 tubes) 
Refrigerated No
Product Line Sorvall
Voltage 120 V