missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Easy Pierce Heat Sealing Foil

Use this heat sealing foil to ready your microplate for compound management and high-throughput screening applications, such as PCR.

$458.06 - $4,551.15


Products 4
Catalog Number Mfr. No. Material Product Type Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Material Product Type Dimensions (L x W) Price Quantity  
AB1720
View Documents
Thermo Scientific™
AB1720
Aluminum EasyPierce 20μm Thin Foil 85 x 135 mm N/A
Only null left
 
AB-0757
View Documents
Thermo Scientific™
AB0757
Aluminum Easy Pierce Foil 85 x 135 mm N/A
Only null left
 
AB-1797
View Documents
Thermo Scientific™
AB1797
Polyester Clear Seal Thin 80 x 125 mm N/A
Only null left
 
AB-0745
View Documents
Thermo Scientific™
AB0745
Aluminum Easy Peel Film Laminate 85 x 135 mm N/A
Only null left
 
Description

Description

Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.

  • Suitable for PCR (requires the pressure from a screw-down heated lid)
  • Easily pierced
  • Peelable bond can be removed by hand
  • 20μm foil is even more easily pierced and is suitable for multiple layering
  • Recommended for use with DMSO
  • Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)

Videos
SDS
Documents

Documents