missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Easy Pierce Heat Sealing Foil
Use this heat sealing foil to ready your microplate for compound management and high-throughput screening applications, such as PCR.
Supplier: Thermo Scientific™ AB1720
Description
Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.
- Suitable for PCR (requires the pressure from a screw-down heated lid)
- Easily pierced
- Peelable bond can be removed by hand
- 20μm foil is even more easily pierced and is suitable for multiple layering
- Recommended for use with DMSO
- Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)
Specifications
| Foil | |
| Aluminum | |
| Not Autoclavable | |
| 85 x 135 mm | |
| General; Long term | |
| Abgene | |
| No | |
| -80°C to +80°C, -80°C to +120°C (with Adequate Pressure) | |
| Polypropylene, Polyethylene | |
| Yes |
| Life Science Reagents Resistant, DMSO Resistant | |
| Non-sterile | |
| 96 | |
| Ideal for compound management and high throughput screening applications, suitable for PCR | |
| EasyPierce 20μm Thin Foil | |
| Polypropylene | |
| Yes | |
| All well shapes | |
| General |