missing translation for 'onlineSavingsMsg'
Learn More

Thermit(R), Granular, Spectrum™ Chemical

$190.71 - $2,381.59

Specifications

Physical Form Solid
Grade Reagent
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614376
View Documents
Spectrum Chemical Mfg Cor
T104625GM
25 g Poly Bottle N/A
Only null left
 
18614377
View Documents
Spectrum Chemical Mfg Cor
T1046500GM
500 g Poly Bottle N/A
Only null left
 
18614374
View Documents
Spectrum Chemical Mfg Cor
T104612KG
12 kg Poly Pail N/A
Only null left
 
Description

Description

Specifications

Specifications

Solid
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.