missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Metalabel™ Series Thermal Transfer Printable Labels, B-435

Printable, metallized polyester labels for thermal transfer printers. Brady™ Thermal Transfer Printable Labels are used for Asset Tracking, Rating Plates, General Purpose ID.

Supplier:  Brady™ THT143510

 Afficher plus de versions de ce produit

Numéro de catalogue. 19-109-205

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Available in various sizes
Spécifications

Spécifications

UL Recognized; CSA Accepted
Acrylic
Gloss
Metallized Polyester (B-435)
Thermal Transfer Printer Label
BBP81;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500
4
THT-1-435-10
10,000/Pk.
19.05 mm (W) x 6.35 mm (H)
-94°F to 266°F
0.0026 in.
8.89 mm
0.75 in.
19.05 mm
6.35 mm
Recommended ribbon series: R4900; R6000; R6200
Silver
Roll
Thermal Transfer
Asset Tracking, Circuit Board Identification - Through Hole Top, General and Industrial Identification, Rating Plates
20.32 mm
-70°C
10000 Label(s) / Roll
Rectangle
19.05 x 6.35 mm (W x H)
-40°C to +90°C
0.0673 mm
6.35 x 19.05 mm
19.05 mm
0.25 in.
THT-1-435-10
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions