missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Thermal Transfer Printable Labels, B-427

Ideal for a variety of applications. Brady™ Thermal Transfer Printable Labels are made of self-laminating vinyl

Supplier:  Brady™ THT1274273

 View more versions of this product

Catalog No. 19-109-197

Only null left
Explore available promotions

Description

Description

  • Available in various sizes
Specifications

Specifications

UL Recognized
Acrylic
Matte
Vinyl, Self-laminating
Thermal Transfer Printer Label
BBP81;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500
-70°C
3000 Label(s) / Roll
Rectangle
55.88 x 12.7 mm (W x H)
-70°C to +70°C
0.102 mm
8 AWG - 1 AWG
2.2 in.
12.70 mm
0.5 in.
12.7 mm
THT-127-427-3
Thermal Transfer Printable Labels
White, Translucent
Roll
Thermal Transfer
General and industrial identification
1
THT-127-427-3
3000/Pk.
12.70 mm (W) x 55.88 mm (H)
-94°F to +158°F
0.004 in.
15.88 mm
55.88 x 12.70 mm
55.88 mm
12.70 mm
0.75 in
19.05 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions