missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ Eam1104I, 3000 U, 150 μL

Catalog No. FERER023T1
Change view
Click to view available options
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
FERER023T1 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERER023T1 Supplier Thermo Scientific™ Supplier No. ER023T1
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.

The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.

Features

  • Convenient stand-alone enzymes for easy workflow optimization
  • Animal-origin-free (AOF) raw materials used in manufacturing
  • No risk of prion or animal virus contamination
  • Reproducible and consistent results with validated quality testing
  • Regulatory compliant—β-lactam-free manufacturing technology
  • Suitable for translational research
  • Facilitates transition to GMP during mRNA vaccine development process
  • Scalable and available as OEM product

The exceptional quality of TheraPure products is supported by:

  • Certificates of analysis
  • Certificates of origin
  • TSE/BSE statement

Specifications

Concentration 20 U/μL
Incubator Temperature 37°C
For Use With (Application) In vitro Transcription
Content And Storage Store at -20°C.
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme Eam1104I
Compatible Buffer 10X Tango Buffer
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE Yes
Quantity 3000 U, 150 μL
Cut Site CTCTTC(1/4)^
Product Type Restriction Enzyme
Research Category Drug Development
Product Line TheraPure
Inactivation Temperature 65°C
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.