Learn More
Therapak™ Refrigerated Biological Substance Category B Shipping Systems
Certified for ground and air shipments of Biological Substance, Category B specimens.
Supplier: Therapak™ 38028
Includes: All systems include transport box with UN3373 Biological Substance, biohazard and orientation arrow labels; EPS foam cooler, gel refrigerant pack(s), 95kPa bags or canisters, a variety of inner packaging choices (depending on the type and and quantity of specimens being transported), absorbent material, list of contents card, cushioning material, and packing instructions.
Description
- DOT and IATA Packing Instruction 650 compliant for the shipment of refrigerated specimens
- Systems suitable for overnight shipments; Mfr. No. 38028 field-tested for 2-8°C temperature maintenance for 48 hours of transport
Chemical Identifiers
| 1 Ea. | |
| DOT and IATA Packing Instruction 650 compliant for the shipment of refrigerated specimens | |
| Refrigerated Shipping for Biological Substance Category B |
Specifications
| 1 Ea. | |
| DOT and IATA Packing Instruction 650 compliant for the shipment of refrigerated specimens | |
| Refrigerated Shipping for Biological Substance Category B | |
| Holds 6 Tubes |
| 95 kPa Bag, Aqui-Pak pouch | |
| For 48 hr. transport | |
| Corrugated outer box (200#, 12 x 12 x 12 in.), EPS cooler with 2.5 in. all-around wall thickness, ambient gel wrap, 95kPa specimen transport bag with absorbent to hold up to 6 specimen tubes, refrigerant packs, list of contents card, filler material and labels for IATA PI650 marking and compliance. | |
| Corrugated box |