Learn More
Therapak™ Refrigerated Biological Substance Category B Shipping Systems
Catalog No. p-2672429
Change view
Click to view available options
Capacity:
Holds 18 Tubes
Holds 6 Tubes
Includes:
95 kPa Bag, Aqui-Pak pouch
95 kPa Canister, 3 Aqui-Pak pouch
95 kPA Canister, Aqui-Pak pouch
Packaging:
Corrugated box
Transport box
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Includes Packaging
22130028 Holds 6 Tubes 95 kPA Canister, Aqui-Pak pouch Transport box
22130032 Holds 18 Tubes 95 kPa Canister, 3 Aqui-Pak pouch Transport box
22130116 Holds 6 Tubes 95 kPa Bag, Aqui-Pak pouch Corrugated box
22130400 Holds 6 Tubes 95 kPa Bag, Aqui-Pak pouch Transport box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22130028 Supplier Therapak™ Supplier No. 37815
Only null left
Includes
All systems include transport box with UN3373 Biological Substance, biohazard and orientation arrow labels; EPS foam cooler, gel refrigerant pack(s), 95kPa bags or canisters, a variety of inner packaging choices (depending on the type and and quantity of specimens being transported), absorbent material, list of contents card, cushioning material, and packing instructions.
Certified for ground and air shipments of Biological Substance, Category B specimens.
  • DOT and IATA Packing Instruction 650 compliant for the shipment of refrigerated specimens
  • Systems suitable for overnight shipments; Mfr. No. 38028 field-tested for 2-8°C temperature maintenance for 48 hours of transport
Specifications
Quantity 1 Ea.
Includes 95 kPA Canister, Aqui-Pak pouch
Certifications/Compliance DOT and IATA Packing Instruction 650 compliant for the shipment of refrigerated specimens
Type Refrigerated Shipping for Biological Substance Category B
Content And Storage Transport box (OD: 11 x 9 x 11 in.), small cooler (ID: 8 x 6 x 8 in.), medium 95kPa canister, Aqui-Pak™ segmented absorbent pouch, 8 oz. refrigerant packs, cushioning material, list of contents card and required labels for IATA PI650 marking and compliance. 
Capacity Holds 6 Tubes
Packaging Transport box