missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Theophylline 98.0+%, TCI America™
Supplier: TCI America T0179100G
Specifications
| Theophylline | |
| 273°C | |
| C7H8N4O2 | |
| 100 g | |
| theophylline, 1,3-dimethylxanthine, elixophyllin, theophyllin, theolair, nuelin, theophylline anhydrous, respbid, theocin, theo-dur | |
| CN1C2=C(C(=O)N(C1=O)C)NC=N2 | |
| 180.167 | |
| CHEBI:28177 | |
| ≥98.0% (HPLC,T) |
| 58-55-9 | |
| White | |
| MFCD00079619 | |
| 1544 | |
| ZFXYFBGIUFBOJW-UHFFFAOYSA-N | |
| 1,3-dimethyl-7H-purine-2,6-dione | |
| 2153 | |
| 180.17 | |
| Crystalline Powder |
Chemical Identifiers
| 58-55-9 | |
| 180.167 | |
| ZFXYFBGIUFBOJW-UHFFFAOYSA-N | |
| 2153 | |
| 1,3-dimethyl-7H-purine-2,6-dione |
| C7H8N4O2 | |
| MFCD00079619 | |
| theophylline, 1,3-dimethylxanthine, elixophyllin, theophyllin, theolair, nuelin, theophylline anhydrous, respbid, theocin, theo-dur | |
| CHEBI:28177 | |
| CN1C2=C(C(=O)N(C1=O)C)NC=N2 |
Safety and Handling
RTECSNumber : XH3850000
TSCA : Yes