missing translation for 'onlineSavingsMsg'
Learn More

Theophylline 98.0+%, TCI America™

Catalog No. p-7136125
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T017925G 25 g
T0179100G 100 g
T0179500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T017925G Supplier TCI America Supplier No. T017925G
Only null left

Chemical Identifiers

CAS 58-55-9
Molecular Formula C7H8N4O2
Molecular Weight (g/mol) 180.167
MDL Number MFCD00079619
InChI Key ZFXYFBGIUFBOJW-UHFFFAOYSA-N
Synonym theophylline, 1,3-dimethylxanthine, elixophyllin, theophyllin, theolair, nuelin, theophylline anhydrous, respbid, theocin, theo-dur
PubChem CID 2153
ChEBI CHEBI:28177
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)NC=N2

Specifications

Melting Point 273°C
Color White
Quantity 25 g
UN Number 1544
Formula Weight 180.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Theophylline
RTECSNumber XH3850000
TSCA Yes