Learn More
Theophylline monohydrate, 99%
CAS: 5967-84-0 | C7H10N4O3 | 198.182 g/mol
Supplier: Thermo Scientific Chemicals A159970B
Description
Theophylline monohydrate is a methylxanthine drug used in therapy for respiratory diseases such as COPD and asthma
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water (8 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| Theophylline monohydrate | |
| 270°C to 274°C | |
| C7H10N4O3 | |
| 1000 g | |
| 13463 | |
| theophylline monohydrate, quibron, quibron plus, theo-sav, dimethylxanthine, aerolate jr, theoclear-100, unii-c137dtr5rg, c7h8n4o2.h2o, theoclear l.a.-260 | |
| INQSMEFCAIHTJG-UHFFFAOYSA-N | |
| 1,3-dimethyl-7H-purine-2,6-dione;hydrate | |
| 91268 | |
| 99% |
| 5967-84-0 | |
| Odorless | |
| MFCD00151659 | |
| UN2811 | |
| 14,9285 | |
| Soluble in water (8g/L at 20°C). | |
| CN1C2=C(C(=O)N(C1=O)C)NC=N2.O | |
| 198.182 | |
| 198.18 (180.16 Anhydrous) |
Chemical Identifiers
| 5967-84-0 | |
| 198.182 | |
| INQSMEFCAIHTJG-UHFFFAOYSA-N | |
| 91268 | |
| CN1C2=C(C(=O)N(C1=O)C)NC=N2.O |
| C7H10N4O3 | |
| MFCD00151659 | |
| theophylline monohydrate, quibron, quibron plus, theo-sav, dimethylxanthine, aerolate jr, theoclear-100, unii-c137dtr5rg, c7h8n4o2.h2o, theoclear l.a.-260 | |
| 1,3-dimethyl-7H-purine-2,6-dione;hydrate |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-385-7
RTECSNumber : XH3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only