CAS RN 5967-84-0
IUPAC Name:
1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione hydrate
Synonyms:
theophylline hydrate
Molecular Weight (g/mol):
198.18
Molecular Formula:
C7H10N4O3
InChi Key:
INQSMEFCAIHTJG-UHFFFAOYSA-N
SMILES:
O.CN1C2=C(NC=N2)C(=O)N(C)C1=O
1
–
1
of
1
results
1
Promotions Available
Theophylline monohydrate, 99%
Theophylline monohydrate, 99%
CAS: 5967-84-0 Molecular Formula: C7H10N4O3 Molecular Weight (g/mol): 198.182 MDL Number: MFCD00151659 InChI Key: INQSMEFCAIHTJG-UHFFFAOYSA-N Synonym: theophylline monohydrate,quibron,quibron plus,theo-sav,dimethylxanthine,aerolate jr,theoclear-100,unii-c137dtr5rg,c7h8n4o2.h2o,theoclear l.a.-260 PubChem CID: 91268 IUPAC Name: 1,3-dimethyl-7H-purine-2,6-dione;hydrate SMILES: CN1C2=C(C(=O)N(C1=O)C)NC=N2.O