missing translation for 'onlineSavingsMsg'
Learn More

Theophylline monohydrate, 99%

Catalog No. AAA1599730
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1599730 250 g
AAA159970B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1599730 Supplier Thermo Scientific Chemicals Supplier No. A1599730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 5967-84-0 | C7H10N4O3 | 198.182 g/mol

Theophylline monohydrate is a methylxanthine drug used in therapy for respiratory diseases such as COPD and asthma

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (8 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5967-84-0
Molecular Formula C7H10N4O3
Molecular Weight (g/mol) 198.182
MDL Number MFCD00151659
InChI Key INQSMEFCAIHTJG-UHFFFAOYSA-N
Synonym theophylline monohydrate, quibron, quibron plus, theo-sav, dimethylxanthine, aerolate jr, theoclear-100, unii-c137dtr5rg, c7h8n4o2.h2o, theoclear l.a.-260
PubChem CID 91268
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione;hydrate
SMILES CN1C2=C(C(=O)N(C1=O)C)NC=N2.O

Specifications

Melting Point 270°C to 274°C
Odor Odorless
Quantity 250 g
UN Number UN2811
Beilstein 13463
Merck Index 14,9285
Solubility Information Soluble in water (8g/L at 20°C).
Formula Weight 198.18 (180.16 Anhydrous)
Percent Purity 99%
Chemical Name or Material Theophylline monohydrate
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-385-7
RTECSNumber XH3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only