missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thaumatin, Thaumatococcus daniellii
A mixture of the intensely sweet proteins thaumatin I and thaumatin II | CAS: 53850-34-3
$144.04 - $692.00
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63668MC
|
Thermo Scientific Chemicals
J63668MC |
100 mg |
Each for $144.04
|
|
|||||
|
AAJ63668MD
|
Thermo Scientific Chemicals
J63668MD |
250 mg |
Each for $278.92
|
|
|||||
|
AAJ6366803
|
Thermo Scientific Chemicals
J6366803 |
1 g |
Each for $692.00
|
|
|||||
Description
Thaumatin is used as a mixture of the intensely sweet proteins, used to enhance the taste of various foods, Flavoring Agents, Food Additives, Sweetening Agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThaumatin is used as a mixture of the intensely sweet proteins, used to enhance the taste of various foods, Flavoring Agents, Food Additives, Sweetening Agents.
Solubility
Soluble in water (25 mg/ml), and 60% ethanol. Insoluble in ether, and benzene .
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
| 53850-34-3 | |
| MFCD00132420 |
Specifications
| 53850-34-3 | |
| MFCD00132420 | |
| 14,9273 | |
| Powder |
| Brown to Yellow | |
| 100 mg | |
| Soluble in water (25mg/ml),and 60% ethanol. Insoluble in ether,and benzene . | |
| Thaumatin, Thaumatococcus daniellii |
Safety and Handling
EINECSNumber : 258-822-2
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only