missing translation for 'onlineSavingsMsg'
Learn More

Acétate thalleux, 99%

Numéro de catalogue. AC420770250
Modifier l'affichage
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Bouteille en verre
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC420770250 25 g Bouteille en verre
AC420771000 100 g Bouteille en verre
AC420775000 500 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC420770250 Fournisseur Thermo Scientific Chemicals Code fournisseur 420770250

CAS : 563-68-8 | C2H3O2Tl | 263,42 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Identifiants chimiques

CAS 563-68-8
Molecular Formula C2H3O2Tl
Molecular Weight (g/mol) 263.42
MDL Number MFCD00013045
InChI Key CNWGLQAFFSLHRX-UHFFFAOYSA-N
Synonym thallium i acetate
PubChem CID 131675083
IUPAC Name acide acétique; l’hydrogène moléculaire; Thallium
SMILES [HH].CC(=O)O.[Tl]

Spécifications

Melting Point 128.0°C to 130.0°C
Color Blanc
Physical Form Poudre fine cristalline
Quantity 25 g
Assay Percent Range 99%
Linear Formula CH3COOTl
Merck Index 15, 9405
Formula Weight 263.42
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Bouteille en verre
Chemical Name or Material Thallous acetate
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-257-5

RUO – Usage réservé à la recherche