accessibility menu, dialog, popup

UserName

CAS RN 563-68-8

IUPAC Name: λ¹-thallanylium acetate
Synonyms: acetic acid, thallium (1+) salt
Molecular Weight (g/mol): 263.42
Molecular Formula: C2H3O2Tl
InChi Key: HQOJMTATBXYHNR-UHFFFAOYSA-M
SMILES: [Tl+].CC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)