missing translation for 'onlineSavingsMsg'
Learn More

Thallium rod, 12.7mm (0.5in) dia, 99.999% (metals basis)

CAS: 7440-28-0 | Tl | 204.38 g/mol

$5486.58 - $5486.58

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
Synonym ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA127140Y
View Documents
Thermo Scientific Chemicals
0127140Y
75 mm
Each for $5,486.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-28-0
204.38
BKVIYDNLLOSFOA-UHFFFAOYSA-N
5359464
thallium
Tl
MFCD00134063
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
CHEBI:30440
[Tl]

Specifications

7440-28-0
Odorless
Rod
MFCD00134063
BKVIYDNLLOSFOA-UHFFFAOYSA-N
thallium
5359464
99.999%
(metals basis)
75 mm
≈225g/150mm
Tl
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
[Tl]
204.38
CHEBI:30440
Thallium rod, 12.7mm (0.5 in.) dia.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

EINECSNumber : 231-138-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only