Learn More
Thallium rod, 12.7mm (0.5in) dia, 99.99% (metals basis)
CAS: 7440-28-0 | Tl | 204.38 g/mol
$3129.89 - $3129.89
Chemical Identifiers
| CAS | 7440-28-0 |
|---|---|
| Molecular Formula | Tl |
| Molecular Weight (g/mol) | 204.38 |
| MDL Number | MFCD00134063 |
| InChI Key | BKVIYDNLLOSFOA-UHFFFAOYSA-N |
| Synonym | ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium |
| PubChem CID | 5359464 |
| ChEBI | CHEBI:30440 |
| IUPAC Name | thallium |
| SMILES | [Tl] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-28-0 | |
| 204.38 | |
| BKVIYDNLLOSFOA-UHFFFAOYSA-N | |
| 5359464 | |
| thallium |
| Tl | |
| MFCD00134063 | |
| ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium | |
| CHEBI:30440 | |
| [Tl] |
Specifications
| 7440-28-0 | |
| Odorless | |
| Rod | |
| MFCD00134063 | |
| BKVIYDNLLOSFOA-UHFFFAOYSA-N | |
| thallium | |
| 5359464 | |
| 99.99% | |
| (metals basis) |
| 75 mm | |
| ≈225g/150mm | |
| Tl | |
| ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium | |
| [Tl] | |
| 204.38 | |
| CHEBI:30440 | |
| Thallium rod, 12.7mm (0.5 in.) dia. |
Safety and Handling
GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
H300+H330-H373
EINECSNumber : 231-138-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only