missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) ethoxide, 95%, Thermo Scientific Chemicals

Catalog No. p-7080349
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA7813106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA7813106 Supplier Thermo Scientific Chemicals Supplier No. 07813106

CAS: 20398-06-5 | Molecular Formula: C2H5OTl | Molecular Weight: 249.441 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20398-06-5
Molecular Formula C2H5OTl
Molecular Weight (g/mol) 249.441
MDL Number MFCD00009072
InChI Key DZFYOYRNBGNPJW-UHFFFAOYSA-N
Synonym thallous ethoxide, thallium i ethanolate, ethoxythallium i, ethanol, thallium 1+ salt, ethyl alcohol, thallium i, ethanol, thallium 1+ salt 1:1, thallium ethoxide, ethanolate; thallium 1+, thallium i ethoxide 5g
PubChem CID 30143
IUPAC Name ethanolate;thallium(1+)
SMILES CC[O-].[Tl+]

Specifications

Boiling Point 130°C (decomposition)
Melting Point -3°C
Physical Form Liquid
Quantity 5 g
Linear Formula TlOCH2CH3
UN Number UN2810
Flash Point >110°C (230°F)
Formula Weight 249.43
Percent Purity 98%
Refractive Index 1.6714
Sensitivity Moisture sensitive
Density 3.493 g/mL
Chemical Name or Material Thallium (I) ethoxide
Show More Show Less
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 243-786-2
RTECSNumber KM2940000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only