accessibility menu, dialog, popup

UserName

CAS RN 20398-06-5

IUPAC Name: λ¹-thallanylium ethanolate
Synonyms: λ¹-thallanylium ethoxide
Molecular Weight (g/mol): 249.44
Molecular Formula: C2H5OTl
InChi Key: DZFYOYRNBGNPJW-UHFFFAOYSA-N
SMILES: [Tl+].CC[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results