missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) acetate, 99.995% (metals basis)

Catalog No. AA1184222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1184222 100 g
AA1184214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1184222 Supplier Thermo Scientific Chemicals Supplier No. 01184222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 563-68-8 | C2H6O2Tl | 266.448 g/mol

Thallium(I) acetate is used as a selective agent against gram-negative bacteria in selective media for the detection of mollicutes. It is also used in preparation of thallium derivatives. It is involved in the separation ore constituents by flotation method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 563-68-8
Molecular Formula C2H6O2Tl
Molecular Weight (g/mol) 266.448
MDL Number MFCD00013045
InChI Key CNWGLQAFFSLHRX-UHFFFAOYSA-N
Synonym thallium i acetate
PubChem CID 131675083
IUPAC Name acetic acid;molecular hydrogen;thallium
SMILES [HH].CC(=O)O.[Tl]

Specifications

Melting Point 131°C
Physical Form Wet Crystalline Aggregates
Quantity 100 g
UN Number UN1707
Beilstein 3693914
Merck Index 14,9257
Solubility Information Soluble in water,alcoholSoluble in cold water,ethanol,chloroform and ether; Sparingly soluble in hot water; Insoluble in acetone.
Formula Weight 263.42
Percent Purity 99.995%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Thallium(I) acetate
Show More Show Less
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
EINECSNumber 209-257-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only