missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged in water
Specifications
Specifications
| Color | Blue-White |
| Quantity | 25 g |
| Odor | Odorless |
| Particle Size | 6mm (0.2 in.) & down |
| UN Number | UN1707 |
| Percent Purity | 99.99% |
| Assay | (metals basis) |
| Chemical Name or Material | Thallium granules |
Safety and Handling
| Hazard Category | H300+H330-H373 |
| Hazard Statement | GHS H Statement H300-H330-H373 Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S. |
| EINECSNumber | 231-138-1 |
| RTECSNumber | XG3425000 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only