missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Thalidomide 98,0+%, TCI America™

Numéro de catalogue. p-7137261
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T25241G 1 g
T25245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T25241G Fournisseur TCI America Code fournisseur T25241G

Identifiants chimiques

CAS 50-35-1
Molecular Formula C13H10N2O4
Molecular Weight (g/mol) 258.23
MDL Number MFCD00153873
InChI Key UEJJHQNACJXSKW-UHFFFAOYNA-N
Synonym thalidomide, thalomid, +/--thalidomide, sedoval, 2-2,6-dioxopiperidin-3-yl isoindoline-1,3-dione, corronarobetin, psycholiquid, psychotablets, theophilcholine, algosediv
PubChem CID 5426
ChEBI CHEBI:74947
IUPAC Name 2-(2,6-dioxopipéridine-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1N(C2CCC(=O)NC2=O)C(=O)C2=CC=CC=C12

Spécifications

Melting Point 276°C
Color Blanc-jaune
Quantity 1 g
UN Number 2811
Formula Weight 258.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Poudre cristalline
Chemical Name or Material (+/-)-Thalidomide
RTECSNumber TI4375000
TSCA No