missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TGM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514966
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514966 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514966 Supplier Invitrogen™ Supplier No. PA514966
Only null left

Rabbit Polyclonal Antibody

TGM2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Transglutaminase (TGase) catalyses the crosslink of proteins by forming e-(g-glutamyl) lysine isopeptide bonds and requires the binding of Ca2+ for its activity. In mammals, eight distinct TGase isoenzymes have been identified. Tissue transglutaminase (tTGase), also known as TGase 2, has four distinct domains: N-terminal b-sandwich, catalytic core and two C-terminal b-barrel domains. tTGase may have a role in cell death, cell proliferation, cell differentiation, and receptor-mediated endocytosis. In the Alzheimer's disease brain, the elevated tTGase activity is manifested by polymerization of a number of proteins, including Ab peptide, b-amyloid precursor protein and the tau protein, with formation of neurofibrillary tangles.

Specifications

Antigen TGM2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TGM2
Gene Accession No. P21980
Gene Alias C polypeptide; C polypeptide, protein-glutamine-gamma-glutamyltransferase; G alpha h; G[a]h; G-ALPHA-h; GNAH; Protein-glutamine gamma-glutamyltransferase 2; protein-glutamine-gamma-glutamyltransferase; RP5-1054A22.2; TG C; TG(C); TG2; TGase C; TGase H; TGase2; TGase-2; TGase-H; TgaseII; TGC; TGM2; Tissue transglutaminase; Tissue type transglutaminase; tissue-type transglutaminase; transglutaminase; transglutaminase 2; transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase); Transglutaminase 2 C polypeptide; transglutaminase 2, C polypeptide; transglutaminase C; Transglutaminase H; transglutaminase-2; tTG; tTGas
Gene Symbols TGM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 429-458 amino acids from the central region of human Transglutaminase (TGM2).
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7052
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less