Learn More
TGF alpha Monoclonal Antibody (MF9), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Thermo Fisher Scientific MA513904
Catalog No. PIMA513904
Only null left
Explore available promotions
Description
Description
MA5-13904 targets Transforming Growth Factor alpha in WB applications and shows reactivity with Bovine, Human, mouse, and Rat samples. The MA5-13904 immunogen is a 17-amino acid synthetic peptide from the carboxyl-terminus of Rat TGFalpha.
TGF-alpha is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-alpha induces anchorage-independence transformation in cultured cells. Human, murine and rat TGF-alpha are cross-species reactive. TGFalpha (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.
Specifications
Specifications
TGF alpha
Monoclonal
0.2 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P01134, P01135, P48030
TGFA
A 17-amino acid synthetic peptide from the carboxyl-terminus of Rat TGFalpha
500 μL
Primary
Bovine, Human, Mouse, Rat
Antibody
IgG1 κ
Western Blot
MF9
Biotin
TGFA
EGF-like TGF; ETGF; I79_019067; LOC100009150; Protransforming growth factor alpha; RATTGFAA; TFGA; TGF a; TGF type 1; TGF a; TGFA; TGFAA; TGFalpha; TGF-alpha; TGFa; Transforming growth factor; Transforming growth factor alpha; transforming growth factor, alpha; transforming growth factor-alpha; wa1; wa-1; waved 1
Mouse
Protein G
RUO
21802, 24827, 540388, 7039
4° C
Liquid
Product Suggestions
Product Suggestions