Learn More
Applied Biosystems™ TFA Aminolink Phosphoramidite, 0.25 g
Catalog No. 402872
Encompass
Change view
Click to view available options
Quantity:
0.25 g
Catalog No. Quantity
402872 0.25 g
1 options
Catalog No. 402872 Supplier Applied Biosystems™ Supplier No. 402872
Each contains 0.25 g TFA Aminolink.TFA Aminolink is used for 5’ attachment of amine group during oligonucleotide synthesis.We offer several phosphoramidites that impart linkers for coupling other labels, and the means to add a phosphate during oligonucleotide synthesis. The label is treated as if it were simply another base: It is inserted at the appropriate place in the sequence and incorporated automatically.
Amine linker:An oligonucleotide incorporating a primary amine readily accepts a variety of labels. TFA Aminolink™ phosphoramidite establishes that amino group at the 5'-end during nucleic acid synthesis.
  • Specialty phosphoramidites allow online addition of labels and linkers during synthesis, saving time in production of labeled probes and primers.
  • They are designed and optimized for use with ABI synthesizers, guaranteeing reliable nucleic acid synthesis.
  • We offer a range of specialty phosphoramites and linkers to give you the flexibility demanded in oligonucleotide synthesis
Specifications
Packaging Type Shipped on Dry Ice
Quantity 0.25 g
For Research Use Only. Not for use in diagnostic procedures.