Learn More
Texwipe™ TexWrite™ 22 Cleanroom Bond Paper
Impregnated with synthetic copolymer that helps reducing risk of particle generation associated with standard papers
Supplier:  Texwipe™ TX5815
 View more versions of this product
Catalog No. 18-344E
Only null left
Explore available promotions
Description
Description
Polymer saturated cleanroom paper reduces particle generation associated with standard paper. Autoclavable for use in sterile environments.
  • Formulated without organic fillers such as calcium carbonate, titanium dioxide or aluminum silicate
  • Possesses both strength and heat resistance, making it ideal for use in laser printers and photocopiers
  • Available in white, blue, yellow, pink and green to allow for easy identification as cleanroom paper or to differentiate between shifts, areas or projects
  • No natural latex binders
  • Excellent toner adhesion
  • Cleanroom packaged
  • Precision-cut edges
Benefits:
  • Very low particle counts and sodium levels
  • Reduced ionic and metal contamination
  • Compatible with toner and offset printing
  • Autoclavable for use in sterile environments
Order Info
Sheets measure 8.5 x 11 in. Packaged 250 sheets per ream, shrink-wrapped and packed in a protective polypropylene bag.
Specifications
Specifications
Synthetic
8.5 x 11 in.
216 mm
279 mm
Standard duty and high speed laser printers and photocopiers, note taking, cleanroom manuals and work instructions
22 lb.
White
8.5 in.
11 in.
Autoclavable
Cleanroom Bond Paper
32.6 gsm
Product Suggestions
Product Suggestions