missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ TexMop™ Aluminum Handles for Cleanroom Mops

For use with AlphaMop™, Mini AlphaMop™/Isolator Cleaning Tool and ClipperMop™.

$552.11 - $552.11

Specifications

Product Type Cleanroom Mop Handle
Color Blue
Quantity 1/Pk.
For Use With (Equipment) AlphaMop, Mini AlphaMop, Isolator Cleaning Tool, ClipperMop
Material Aluminum
View More Specs

Products 1
Catalog Number Mfr. No. Length (English) Price Quantity  
Catalog Number Mfr. No. Length (English) Price Quantity  
18-366-257
View Documents
Texwipe™
TX7183
55 to 92 in.
Each for $552.11
Only null left
 
Description

Description

  • Telescoping type
  • Blue color
  • Use with TexConnect™ adapter to attach to screw-based mop heads
Specifications

Specifications

Cleanroom Mop Handle
1/Pk.
Aluminum
Blue
AlphaMop, Mini AlphaMop, Isolator Cleaning Tool, ClipperMop
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'