missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Texas Red™ Sulfonyl Chloride, mixed isomers

Catalog No. t353
Encompass
Change view
Click to view available options
Quantity:
10 mg
10 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T353 10 mg
T1905 10 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T353 Supplier Invitrogen™ Supplier No. T353

Texas Red™ Sulfonyl Chloride, mixed isomers

The amine-reactive Texas Red™ sulfonyl chloride can be used to can be used to create bright red-fluorescent bioconjugates with excitation/emission maxima ∼595/615 nm.

Specifications

Quantity 10 mg
Product Type Sulfonyl Chloride
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Sulfonyl Chloride
Label or Dye Texas Red™
Label Type Classic Dyes
Product Line Texas Red
Emission 615
Excitation 595
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.