missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrolic Acid 98.0+%, TCI America™
$70.84 - $848.14
Chemical Identifiers
| CAS | 590-93-2 |
|---|---|
| Molecular Formula | C4H4O2 |
| Molecular Weight (g/mol) | 84.07 |
| MDL Number | MFCD00004363 |
| InChI Key | LUEHNHVFDCZTGL-UHFFFAOYSA-N |
| Synonym | 2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid |
| PubChem CID | 68535 |
| IUPAC Name | but-2-ynoic acid |
| SMILES | CC#CC(O)=O |
Chemical Identifiers
| 590-93-2 | |
| 84.07 | |
| LUEHNHVFDCZTGL-UHFFFAOYSA-N | |
| 68535 | |
| CC#CC(O)=O |
| C4H4O2 | |
| MFCD00004363 | |
| 2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid | |
| but-2-ynoic acid |
Specifications
| 590-93-2 | |
| White-Yellow | |
| C4H4O2 | |
| 1 g | |
| 2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid | |
| CC#CC(O)=O | |
| 84.07 | |
| 84.07 | |
| Crystalline Powder |
| 77°C | |
| 203°C | |
| MFCD00004363 | |
| 3261 | |
| LUEHNHVFDCZTGL-UHFFFAOYSA-N | |
| but-2-ynoic acid | |
| 68535 | |
| ≥98.0% (GC,T) | |
| Tetrolic Acid |
Safety and Handling
TSCA : Yes